Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NoTrax™ Pebble Trax™
GREENER_CHOICE

Catalog No. 19085912 Shop All NoTrax Products
Change view
Click to view available options
Dimensions (L x W):
2 x 3 ft.
3 x 5 ft.
3 x 12 ft.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-085-912 2 x 3 ft.
19-085-913 3 x 5 ft.
19-085-914 3 x 12 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-085-912 Supplier NoTrax™ Supplier No. 480S0023BL
Add to Cart
Add to Cart

Pebble Trax™ is an anti-fatigue floor mat for assembly lines, lab areas, machine workstations and packing areas

  • Anti-fatigue floor mat offers comfort and ergonomic support
  • For dry environments like assembly lines, lab areas, machine workstations and packing areas
  • Pebble-embossed rubber top provides non-directional traction
  • Easy to clean
  • Resists exposure to sparks from welding
  • Made with a resilient SBR rubber compound, top surface combined with dense closed cell foam base utilizing Notrax™ exclusive UniFusion™ technology
    • Eliminates the possibility of de-lamination
  • Comes standard with RedStop™
    • Uniquely engineered non-slip backing technology eliminates slipping and sliding of floor mats on smooth surface flooring such as tile, wood, marble, and treated concrete
  • Sloped beveled edges reduce tripping hazards and allow easy access on and off mat

Specifications

Type Anti-fatigue Floor Mat
Dimensions (L x W) 2 x 3 ft.
Length (English) 2 ft.
Width (English) 3 ft.
Color Black, Yellow
Material Top Surface Dense Closed Cell PVC Foam Base
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.