Learn More
MSC PEC Tools Combination Square Blade, 18 in, 16R, Hardened, Satin Chrome
Catalog No. 5000533191
Encompass
This combination square blade is a high-precision measuring tool designed for engineers, machinists, and craftsmen. Crafted from quality steel, this individual hardened blade features a satin chrome finish and precisely etched, black-filled graduations for excellent readability and durability. It is designed for use as a replacement or component for a combination square.
  • All blades fully hardened and tempered.
  • Satin chrome finish for durability.
  • Made from the finest quality steel.
  • Precisely etched and black-filled for readability.
  • Features 16R graduations (32nds, 64ths, 50ths, 100ths).
Catalog No. 50-005-33191 Supplier MSC Supplier No. 06505036
This combination square blade is a high-precision measuring tool designed for engineers, machinists, and craftsmen. Crafted from quality steel, this individual hardened blade features a satin chrome finish and precisely etched, black-filled graduations for excellent readability and durability. It is designed for use as a replacement or component for a combination square.
  • All blades fully hardened and tempered.
  • Satin chrome finish for durability.
  • Made from the finest quality steel.
  • Precisely etched and black-filled for readability.
  • Features 16R graduations (32nds, 64ths, 50ths, 100ths).