Learn More
MSC PEC Tools Drill Gauge, Industrial Quality, Hardened, 61-80 Wire Size
Catalog No. 5000534266
Encompass
This Industrial Quality - Hardened Twist Drill and Drill Rod Gauge from PEC Tools is designed for precision measurement of wire sizes ranging from 61 to 80. Its robust construction ensures durability and accuracy for various industrial applications.
  • Industrial quality construction.
  • Hardened for enhanced durability.
  • Designed for twist drills and drill rods.
  • Measures wire sizes 61 to 80.
Catalog No. 50-005-34266 Supplier MSC Supplier No. 01471093
This Industrial Quality - Hardened Twist Drill and Drill Rod Gauge from PEC Tools is designed for precision measurement of wire sizes ranging from 61 to 80. Its robust construction ensures durability and accuracy for various industrial applications.
  • Industrial quality construction.
  • Hardened for enhanced durability.
  • Designed for twist drills and drill rods.
  • Measures wire sizes 61 to 80.