Learn More
MSC PEC Tools Inside Caliper, 12 in, Solid Nut
Catalog No. 5000529946
Encompass
PEC Tools inside calipers are designed for precise internal measurements. They feature balanced legs for uniform tension and easy adjustment across their entire range, making them suitable for various measuring tasks.
  • Balanced legs, uniform tension
  • Polished natural steel finish
  • Easy adjustment over entire range
  • Centrally located adjustment screw
Catalog No. 50-005-29946 Supplier MSC Supplier No. 04968467
PEC Tools inside calipers are designed for precise internal measurements. They feature balanced legs for uniform tension and easy adjustment across their entire range, making them suitable for various measuring tasks.
  • Balanced legs, uniform tension
  • Polished natural steel finish
  • Easy adjustment over entire range
  • Centrally located adjustment screw