Learn More
MSC PEC Tools Outside Caliper, Quick Nut, 6 in, Steel
Catalog No. 5000510925
Encompass
This outside caliper features balanced legs, uniform tension, and a polished natural steel finish. It allows for easy adjustment over its entire range with a centrally located adjustment screw, and this specific model utilizes a Quick Nut for precise and efficient adjustments.
  • Balanced legs and uniform tension
  • Polished natural steel finish
  • Easy adjustment over entire range
  • Centrally located adjustment screw
  • Quick Nut for adjustment
Catalog No. 50-005-10925 Supplier MSC Supplier No. 83306076
This outside caliper features balanced legs, uniform tension, and a polished natural steel finish. It allows for easy adjustment over its entire range with a centrally located adjustment screw, and this specific model utilizes a Quick Nut for precise and efficient adjustments.
  • Balanced legs and uniform tension
  • Polished natural steel finish
  • Easy adjustment over entire range
  • Centrally located adjustment screw
  • Quick Nut for adjustment