Learn More
MSC PEC Tools Protractor and Depth Gage, 6 Inch Rule, 180° Angle
Catalog No. 5000510719
Encompass
This protractor and depth gage offers precise measurements with a 0 to 6 inch rule range and a 0 to 180 degree angle range. It is constructed with a durable tempered steel, featuring a rectangle head for stability.
  • 0 to 6 inch rule measurement range
  • 0 to 180 degree angle measurement range
  • Rectangle head for stable measurements
  • Constructed from tempered steel for durability
  • Angle graduation of 1 degree
  • Head length of 3-5/16 inch
  • Head width of 2 inch
  • Manufactured in the United States
Catalog No. 50-005-10719 Supplier MSC Supplier No. 04930236
This protractor and depth gage offers precise measurements with a 0 to 6 inch rule range and a 0 to 180 degree angle range. It is constructed with a durable tempered steel, featuring a rectangle head for stability.
  • 0 to 6 inch rule measurement range
  • 0 to 180 degree angle measurement range
  • Rectangle head for stable measurements
  • Constructed from tempered steel for durability
  • Angle graduation of 1 degree
  • Head length of 3-5/16 inch
  • Head width of 2 inch
  • Manufactured in the United States