Learn More
MSC PEC Tools Steel Rule, 6 inch, 4R, Rigid, Satin Chrome-Plated
Catalog No. 5000528698
Encompass
This rigid steel rule is made from the finest quality steel, fully hardened and tempered for durability. It features a baked-on satin chrome finish for enhanced readability and corrosion prevention. The precision photo etched and black-filled graduations ensure accurate measurements.
  • Made from finest quality steel
  • Fully hardened and tempered for durability
  • Precisely etched and black-filled graduations for readability
  • Satin chrome finish for easy readability and corrosion prevention
  • Rigid construction
  • Six inch length
  • 4R graduations
  • Both long edges ground flat and straight
  • Made in USA
Catalog No. 50-005-28698 Supplier MSC Supplier No. 80559792
This rigid steel rule is made from the finest quality steel, fully hardened and tempered for durability. It features a baked-on satin chrome finish for enhanced readability and corrosion prevention. The precision photo etched and black-filled graduations ensure accurate measurements.
  • Made from finest quality steel
  • Fully hardened and tempered for durability
  • Precisely etched and black-filled graduations for readability
  • Satin chrome finish for easy readability and corrosion prevention
  • Rigid construction
  • Six inch length
  • 4R graduations
  • Both long edges ground flat and straight
  • Made in USA