Learn More
MSC PEC Tools Steel Rule, 6 Inch, 5R Graduation, Rigid, Chrome Finish
Catalog No. 5000527839
Encompass
This steel rule is designed for precise measurements in various applications. It features a rigid steel construction with a durable chrome finish, offering clear and accurate readings.
  • 6 inch length
  • 5R graduation style
  • Graduations include 1/64, 1/100, and 1/32 inch
  • Rigid design for stable measurements
  • Steel material ensures durability
  • Chrome finish for enhanced readability and protection
  • 3/4 inch width
  • 0.0350 inch thickness
Catalog No. 50-005-27839 Supplier MSC Supplier No. 06511422
This steel rule is designed for precise measurements in various applications. It features a rigid steel construction with a durable chrome finish, offering clear and accurate readings.
  • 6 inch length
  • 5R graduation style
  • Graduations include 1/64, 1/100, and 1/32 inch
  • Rigid design for stable measurements
  • Steel material ensures durability
  • Chrome finish for enhanced readability and protection
  • 3/4 inch width
  • 0.0350 inch thickness