Learn More
MSC PEC Tools Steel Rule, Steel, 6 in, English & Metric Graduation, Rigid
Catalog No. 5000529839
Encompass
This steel rule is a rigid measuring instrument designed for precision. It features both English and Metric graduations, making it versatile for various applications. The rule is made from durable steel and finished with satin chrome for enhanced readability and corrosion resistance.
  • Constructed from durable steel.
  • Measures 6 inches in overall length.
  • Features both English and Metric graduation styles.
  • Rigid design ensures stable and accurate measurements.
  • Satin chrome finish provides improved readability and corrosion resistance.
  • Has a width of 3/4 "
  • Features a thickness of 0.0350 "
Catalog No. 50-005-29839 Supplier MSC Supplier No. 01470905
Add to Cart
Add to Cart
This steel rule is a rigid measuring instrument designed for precision. It features both English and Metric graduations, making it versatile for various applications. The rule is made from durable steel and finished with satin chrome for enhanced readability and corrosion resistance.
  • Constructed from durable steel.
  • Measures 6 inches in overall length.
  • Features both English and Metric graduation styles.
  • Rigid design ensures stable and accurate measurements.
  • Satin chrome finish provides improved readability and corrosion resistance.
  • Has a width of 3/4 "
  • Features a thickness of 0.0350 "