Learn More
Electron Microscopy Sciences Peca Techni-Valve
SDP
Catalog No. 50930503
Encompass_Preferred
For use with our Dust-Pro Refill cans. Professionally designed with a long nozzle (3 1/2",90 mm). Smooth bell housing and no mounting clip is required. Made from solid brass, chrome plated. The valve can be removed at any time without the loss of pressure. Patented bell housing fits the contour of your hand.
Catalog No. 50-930-503 Supplier Electron Microscopy Sciences Supplier No. 7084910
For use with our Dust-Pro Refill cans. Professionally designed with a long nozzle (3 1/2",90 mm). Smooth bell housing and no mounting clip is required. Made from solid brass, chrome plated. The valve can be removed at any time without the loss of pressure. Patented bell housing fits the contour of your hand.