Learn More
Aviva Systems Biology Corp PECI Antibody - middle region (ARP48692_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48692_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PECI. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PECI. Alternate names: ACBD2, DRS1, HCA88, dJ1013A10.3, PECI, DRS-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59886