Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pectin, Citrus, USP, Spectrum™ Chemical
SDP

Catalog No. 18604999 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
20 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-999 20 kg Poly Bottle
18-604-997 500 g Poly Bottle
18-604-998 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-604-999 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PE10020KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PE100, 9000-69-5

Spectrum™ Chemical Pectin, Citrus, USP is used as a gelling agent and thickening agent. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9000-69-5

Specifications

CAS Max % 1
Quantity 20 kg
Loss on Drying 0.1
Packaging Poly Bottle
Grade USP
Chemical Name or Material Pectin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.