Learn More
Sigma Aldrich Fine Chemicals Biosciences Pectin from citrus peel, 9000-69-5, MFCD00081838, 100 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P9135100G
Encompass
Galacturonic acid ≥74.0 % (dried basis), storage temp: room temp, Synonym: Poly-D-galacturonic acid methyl ester. Pectin from citrus peel has been used: in the comparative study of physicochemical, rheological and gelling properties of pectin among different extraction method, in starch suspension for measuring in vitro starch digestibility, to measure pectinase activity, and in the production of pectinase on solid medium.
Catalog No. NC1045358