Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pectin, MP Biomedicals™

Catalog No. ICN10258790 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10258790 500 g
ICN10258780 100 g
ICN10258791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10258790 Supplier MP Biomedicals, Inc Supplier No. 0210258790
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9000-69-5
Molecular Formula (C7H10O7)n
Synonym Poly-D-galacturonic acid methyl ester

Specifications

Color Beige to White
Physical Form Powder
Quantity 500 g
Formula Weight 20,000 to 400,000
Chemical Name or Material Pectin
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.