Learn More
Bioworld Pectinase, 10 g
Catalog No. 50198987
Encompass_Preferred
Pectinase Also known as Poly-(1,4-a-D-galacturonide)glycanohydrolase. Used in plant protoplast preparation to digest cell wall prior to organelle isolation.
Catalog No. 50-198-987 Supplier Bioworld Supplier No. 215600192
Pectinase Also known as Poly-(1,4-a-D-galacturonide)glycanohydrolase. Used in plant protoplast preparation to digest cell wall prior to organelle isolation.