Learn More
Bioworld Pectinase, 1 g
Catalog No. 50198986
Encompass_Preferred
Pectinase Also known as Poly-(1,4-a-D-galacturonide)glycanohydrolase. Used in plant protoplast preparation to digest cell wall prior to organelle isolation.
Catalog No. 50-198-986 Supplier Bioworld Supplier No. 215600191
Pectinase Also known as Poly-(1,4-a-D-galacturonide)glycanohydrolase. Used in plant protoplast preparation to digest cell wall prior to organelle isolation.