Learn More
Sigma Aldrich Fine Chemicals Biosciences Pectinase from Aspergillus aculeatus | 50 mL
Catalog No. NC1697623
Encompass
Pectinase is an enzyme from Aspergillus aculeatus that is used in plant protoplast preparation to digest cell wall prior to organelle isolation. It has been used to conduct partial saccharification of sugars. Pectinases are used to study their role in the invasion of plant tissues by phytopathogens, the spoilage of produce and various food processing and plant biotechnology applications. Pectinase, product P2611, has been used to treat leaf discs from fifth wild-type and sitiens leaves.
Catalog No. NC1697623 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P261150ML
Pectinase is an enzyme from Aspergillus aculeatus that is used in plant protoplast preparation to digest cell wall prior to organelle isolation. It has been used to conduct partial saccharification of sugars. Pectinases are used to study their role in the invasion of plant tissues by phytopathogens, the spoilage of produce and various food processing and plant biotechnology applications. Pectinase, product P2611, has been used to treat leaf discs from fifth wild-type and sitiens leaves.