Learn More
Medchemexpress LLC Pectinase, aspergillus niger | 9032-75-1 | 10 G
SDP
Catalog No. 5000438075
Encompass_Preferred
Pectinase (EC 3.2.1.15) is a mixed enzyme that hydrolyzes pectic substances and is primarily found in microorganisms and higher plants. It plays a role in cell wall metabolism, cell growth, senescence, fruit ripening, pathogenesis, and the abscission process. This product is derived from Aspergillus niger fermentation.
  • Effective temperature range 25-65°C
  • Optimal operating temperature 50-60°C
  • Effective pH range 2.5-4.5
  • Optimal pH 3.3-4.0
  • Activators: Ca2+, Mg2+
  • Inhibitors: Cu2+, Fe3+
  • Appearance: Solid, light yellow to light brown
  • Insoluble in H2O (< 0.1 mg/mL)
Catalog No. 50-004-38075 Supplier Medchemexpress LLC Supplier No. HY12638610G
Add to Cart
Add to Cart
Pectinase (EC 3.2.1.15) is a mixed enzyme that hydrolyzes pectic substances and is primarily found in microorganisms and higher plants. It plays a role in cell wall metabolism, cell growth, senescence, fruit ripening, pathogenesis, and the abscission process. This product is derived from Aspergillus niger fermentation.
  • Effective temperature range 25-65°C
  • Optimal operating temperature 50-60°C
  • Effective pH range 2.5-4.5
  • Optimal pH 3.3-4.0
  • Activators: Ca2+, Mg2+
  • Inhibitors: Cu2+, Fe3+
  • Appearance: Solid, light yellow to light brown
  • Insoluble in H2O (< 0.1 mg/mL)