Learn More
Gold Biotechnology Inc Pectolyase Y-23 1 g
SDP
Catalog No. NC2007207
Encompass
Pectolyase Y-23 is a highly purified maceration enzyme from Aspergillus japonicus. It contains two types of pectinase such as endo-polygalacturonase and endo-pectin lyase in high activity.
Catalog No. NC2007207 Supplier Gold Biotechnology Inc Supplier No. P8004.0001
Pectolyase Y-23 is a highly purified maceration enzyme from Aspergillus japonicus. It contains two types of pectinase such as endo-polygalacturonase and endo-pectin lyase in high activity.