Learn More
Abcam PE/Cy5.5 Conjugation Kit - Lightning-Link, 600UG
Catalog No. 502585592
Encompass_Preferred
Labeling in < 4 hrs with 30 secs hands-on time using PE/Cy5.5 Conjugation Kit - Lightning-Link ab102899. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5592 Supplier Abcam Supplier No. AB102899600UG
Labeling in < 4 hrs with 30 secs hands-on time using PE/Cy5.5 Conjugation Kit - Lightning-Link ab102899. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement