Learn More
Abcam PE/Cy5 Conjugation Kit - Lightning-Link, 3 x 60UG
Catalog No. 502585596
Encompass_Preferred
The Abcam PE/Cy5 Conjugation Kit ( ab102893) offers several standout features: - Rapid Conjugation: Achieve PE/Cy5 labeling in under 4 hours with just 30 seconds of hands-on time. - High Efficiency: Ensures 100% antibody recovery, preserving the integrity and value of your antibodies. - Versatility: Suitable for conjugating antibodies, proteins, and peptides, and the labeled antibodies can be used immediately in applications such as Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5596 Supplier Abcam Supplier No. AB1028933X60UG
The Abcam PE/Cy5 Conjugation Kit ( ab102893) offers several standout features: - Rapid Conjugation: Achieve PE/Cy5 labeling in under 4 hours with just 30 seconds of hands-on time. - High Efficiency: Ensures 100% antibody recovery, preserving the integrity and value of your antibodies. - Versatility: Suitable for conjugating antibodies, proteins, and peptides, and the labeled antibodies can be used immediately in applications such as Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification.
The product is subject to the following: Abcam Restricted Use Statement