Learn More
Biolegend PE/Cyanine7 anti-human CD133
Supplier:  Biolegend 394110
Encompass_Preferred
PE/Cyanine7 anti-human CD133 [S16016E]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-9095