Learn More
Biolegend PE/Cyanine7 anti-human CD279 (PD-1)
Supplier:  Biolegend 621615
Encompass_Preferred
PE/Cyanine7 anti-human CD279 (PD-1) [A17188B]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-208-6792