Learn More
Biolegend PE/Cyanine7 anti-human CD66b Recombinant
Supplier:  Biolegend 396909
Encompass_Preferred
PE/Cyanine7 anti-human CD66b Recombinant [QA17A51]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-9262