Learn More
Biolegend PE/Dazzle™ 594 anti-human CD340 (erbB2/HER-2)
Supplier:  Biolegend 324430
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD340 (erbB2/HER-2) [24D2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-5666