Learn More
Bellco Glass, Inc. Pedestal Stand, 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800452
Encompass_Preferred
Hands-Free Media Filtering Made Easy and AffordableUse in Conjunction with the available separately Autofil filtration system and/or the Cradle Ring. These two filtering options offer a permanent vacuum hose connection and minimize set-up time. The vacuum hose connects to the back of the pedestal or cradle ring and enables adjustable flow control using the on/off switch. This patented docking system is made from aluminum.Patented docking hardware:Allows for hands-free filtrationProvides on/off vacuum flow controlPermanent hose connection to minimize set-up timeWorks in conjunction with Autofil system in 15ml, 50ml, 250ml, 500ml, and 1000mlSPECIFICATIONSColor: BlackMade in: USAMaterial: AluminumSize/Dimensions;Overall height: 8"Base diameter: 7"Base thickness: 1.5"Clearance - base to dock: 5.75"
Catalog No. 50-255-0310