Learn More
Med Vet International Pediatric "F" Anesthesia Circuit, 60" Hose w/o Bag, J0248KLA
Catalog No. 5000582222
Encompass_Preferred
For animals weighing 4Kg–20Kg.Features: Smaller diameter tubing with less dead space than standard uni-limb system Pink colored inner tubing 40" one 60" hose
Catalog No. 50-005-82222 Supplier Med Vet International Supplier No. JOR248KLA
For animals weighing 4Kg–20Kg.Features: Smaller diameter tubing with less dead space than standard uni-limb system Pink colored inner tubing 40" one 60" hose