Learn More
Med Vet International Pediatric Universal "F" Anesthesia Circuit With 1L Bag, J0248K
Catalog No. 5000582369
Encompass_Preferred
For smaller animals weighing between 4kg and 20kg. Includes a smaller diameter tubing with less dead space than the standard uni-limb system. Pink inner tubing.
Catalog No. 50-005-82369 Supplier Med Vet International Supplier No. JOR248K
For smaller animals weighing between 4kg and 20kg. Includes a smaller diameter tubing with less dead space than the standard uni-limb system. Pink inner tubing.