Learn More
Biolegend PE/Dazzle™ 594 anti-human CD105
Supplier:  Biolegend 323223
Encompass_Preferred
PE/Dazzle™ 594 anti-human CD105 [43A3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-5594