Learn More
Analytical Sales & Services PEEK 2 Position, 6 Port Analytical Syringe Load Manual Inert Sample Injector with Attached 20µL Sample Loop
Catalog No. 5000807045
Encompass
PEEK 2 Position 6 Port Analytical Syringe Load Manual Inert Sample Injector with Attached 20µL Sample Loop
Catalog No. 50-008-07045 Supplier Analytical Sales & Services Supplier No. 497254
PEEK 2 Position 6 Port Analytical Syringe Load Manual Inert Sample Injector with Attached 20µL Sample Loop