Learn More
Analytical Sales & Services PEEK 2 Position, 6 Port Analytical Syringe Load Manual Inert Sample Injector with Built In Position Sensing Switch, Attached 20µL Sample Loop
Catalog No. 5000804917
Encompass
PEEK 2 Position, 6 Port Standard Analytical Syringe Load Manual Inert Sample Injector with Built In Position Sensing Switch, Attached 20µL Sample Loop, 5,000psi (345 bar)
Catalog No. 50-008-04917 Supplier Analytical Sales & Services Supplier No. 49725I4
PEEK 2 Position, 6 Port Standard Analytical Syringe Load Manual Inert Sample Injector with Built In Position Sensing Switch, Attached 20µL Sample Loop, 5,000psi (345 bar)