Learn More
Analytical Sales & Services PEEK 2 Position, 6 Port High Pressure Switching Valve
Catalog No. 5000805700
Encompass
PEEK 2 Position, 6 Port High Pressure Switching Valve. 5,000psi (345 bar)
Qty/Pkg: EACH
Catalog No. 50-008-05700 Supplier Analytical Sales & Services Supplier No. VXV7788000
PEEK 2 Position, 6 Port High Pressure Switching Valve. 5,000psi (345 bar)
Qty/Pkg: EACH