Learn More
Analytical Sales & Services PEEK 2 Position, 6 Port Prep Syringe Load Manual Sample Injector with Built In Position Sensing Switch
Catalog No. 5000804221
Encompass
PEEK 2 Position, 6 Port Prep Syringe Load Manual Sample Injector with Built In Position Sensing Switch and Attached 10mL Sample Loop.\nInject from 10 - 100mL/min. Set for 3,000psi. Adjustable to 4,000psi.\n\nQty/Pkg: EACH
Catalog No. 50-008-04221 Supplier Analytical Sales & Services Supplier No. 43725I4
PEEK 2 Position, 6 Port Prep Syringe Load Manual Sample Injector with Built In Position Sensing Switch and Attached 10mL Sample Loop.\nInject from 10 - 100mL/min. Set for 3,000psi. Adjustable to 4,000psi.\n\nQty/Pkg: EACH