Learn More
IDEX Health & Science Upchurch PEEK ASSY 1/8 NPT-1/4-28 FEM.
Catalog No. NC1919893
Encompass_Preferred
Idex National Pipe Thread (NPT) Adapters, For Use With: 0.125 in. Male NPT to 0.25-28 Female, Includes: P-300/308 for 0.125 in. O.D. tubing, PEEK, NPT Adapter, Red, Diameter Inner: 0.062 in.
Catalog No. NC1919893 Supplier IDEX Health & Science Upchurch Supplier No. U510
Idex National Pipe Thread (NPT) Adapters, For Use With: 0.125 in. Male NPT to 0.25-28 Female, Includes: P-300/308 for 0.125 in. O.D. tubing, PEEK, NPT Adapter, Red, Diameter Inner: 0.062 in.