Learn More
Analytical Sales & Services PEEK Column Coupler
Catalog No. 5000806603
Encompass
Advantage PEEK 2 Piece Column Coupler Includes Two Pair of 2- Piece Fingertight Fittings and One Length of 0.01"ID PEEK Tubing.
Qty/Pkg: EACH
Catalog No. 50-008-06603 Supplier Analytical Sales & Services Supplier No. 64287
Advantage PEEK 2 Piece Column Coupler Includes Two Pair of 2- Piece Fingertight Fittings and One Length of 0.01"ID PEEK Tubing.
Qty/Pkg: EACH