Learn More
Microsolv Technology Corporation HPLC Column Set, PEEK, 2.1mm ID. Includes, 1 EA 250mm x 2.1mm Column Tube, Black, 2 EA 2um Frits, 2 EA End Fittings. 1 EA.
Catalog No. 502271600
Encompass_Preferred
This complete column set is made with Victrex PEEK and we add graphite to achieve the black color of the 1/4" OD Column Tube, which is not treated with any chemicals. The polished interior is achieved through a very careful, proprietary, extrusion method.
This item is not returnable. View return policy
Catalog No. 50-227-1600 Supplier Microsolv Technology Corporation Supplier No. 49334250
This complete column set is made with Victrex PEEK and we add graphite to achieve the black color of the 1/4" OD Column Tube, which is not treated with any chemicals. The polished interior is achieved through a very careful, proprietary, extrusion method.