Learn More
Microsolv Technology Corporation HPLC Column Set, PEEK, 7.5mm ID. Includes, 1 EA 150mm x 7.5mm Column Tube, Black, 2 EA 5um Frits, 2 EA End Fittings. 1 EA.
Catalog No. 502271566
Encompass_Preferred
This complete column set is made with Victrex PEEK and we add graphite to achieve the black color of the 5/8" OD Column Tube, which is not treated with any chemicals. The polished interior is achieved through a very careful, proprietary, extrusion method.
This item is not returnable. View return policy
Catalog No. 50-227-1566 Supplier Microsolv Technology Corporation Supplier No. 49333150
This complete column set is made with Victrex PEEK and we add graphite to achieve the black color of the 5/8" OD Column Tube, which is not treated with any chemicals. The polished interior is achieved through a very careful, proprietary, extrusion method.