Learn More
Analytical Sales & Services PEEK Cross for 1/16" OD Tubing
Catalog No. 5000805057
Encompass
PEEK Cross with .020" Thru-Hole for 1/16" OD Tubing\n\nQty/Pkg: EACH
Catalog No. 50-008-05057 Supplier Analytical Sales & Services Supplier No. 59722
Add to Cart
Add to Cart
PEEK Cross with .020" Thru-Hole for 1/16" OD Tubing\n\nQty/Pkg: EACH