Learn More
Analytical Sales & Services PEEK Female Nut, 5/16-24
Catalog No. 5000805077
Encompass
PEEK Female Nut for Micro Tees, Micro Crosses and MicroFilter Assemblies\n• 5/16-24 Internal Threads\n\nQty/Pkg: EACH
Catalog No. 50-008-05077 Supplier Analytical Sales & Services Supplier No. 59416
PEEK Female Nut for Micro Tees, Micro Crosses and MicroFilter Assemblies\n• 5/16-24 Internal Threads\n\nQty/Pkg: EACH