Learn More
Analytical Sales & Services PEEK Fitting for Microsleeves
Catalog No. 5000804658
Encompass
PEEK Micro Standard Head Fingertight Fittings for Microsleeves\n\nThreads: 6-32\n\nQty/Pkg: Each
Catalog No. 50-008-04658 Supplier Analytical Sales & Services Supplier No. 55125
PEEK Micro Standard Head Fingertight Fittings for Microsleeves\n\nThreads: 6-32\n\nQty/Pkg: Each