Learn More
Waters Corp PEEK Fitting, One Piece, Short
Supplier:  Waters Corp 700002842
Encompass_Preferred
This PEEK fitting is a one-piece, short component used in chromatography systems. It is designed to replace a micro-union when a new emitter tip is being used, particularly with the Universal NanoFlow sprayer. The fitting allows for fingertight installation, securing the emitter in the micro-union.
  • One-piece design
  • Short component for specific applications
  • Used with Universal NanoFlow sprayer
  • Secures emitter in micro-union
  • Fingertight installation for ease of use
  • Replaces micro-union for new emitter tips
Catalog No. NC3852720