Learn More
Analytical Sales & Services PEEK Fittings for Microsleeves
Catalog No. 5000804506
Encompass
PEEK Micro Standard Head Fingertight Fittings for Microsleeves\n\nThreads: 6-32\n\nQty/Pkg: 10
Catalog No. 50-008-04506 Supplier Analytical Sales & Services Supplier No. 55125X
PEEK Micro Standard Head Fingertight Fittings for Microsleeves\n\nThreads: 6-32\n\nQty/Pkg: 10