Learn More
Waters Corp WATERS CORPORATION PEEK Fittings, Double Ferrule, 1/16 in., 10-32 Thread
Catalog No. 50784245 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product consists of PEEK fittings equipped with double ferrules, designed for secure connections in systems requiring inert and biocompatible components. The fittings are specified for 1/16 in. tubing and feature a 10-32 thread with a length of 1/2 in. or 0.525 in.
  • Made from PEEK material
  • Includes double ferrules for enhanced sealing
  • Designed for 1/16 in. tubing
  • Utilizes a 10-32 thread
Catalog No. 50-784-245 Supplier Waters Corp Supplier No. PSL613325
This product consists of PEEK fittings equipped with double ferrules, designed for secure connections in systems requiring inert and biocompatible components. The fittings are specified for 1/16 in. tubing and feature a 10-32 thread with a length of 1/2 in. or 0.525 in.
  • Made from PEEK material
  • Includes double ferrules for enhanced sealing
  • Designed for 1/16 in. tubing
  • Utilizes a 10-32 thread