Learn More
Analytical Sales & Services PEEK Flangeless Nut for 1/16" OD Tubing
Catalog No. 5000805090
Encompass
Natural PEEK Flangeless nut, 5/16-24 Flat-Bottom
Catalog No. 50-008-05090 Supplier Analytical Sales & Services Supplier No. 59136
Natural PEEK Flangeless nut, 5/16-24 Flat-Bottom