Learn More
Analytical Sales & Services PEEK Flangeless Nut for 1/16" OD Tubing
Catalog No. 5000804598
Encompass
1/4-28 PEEK Flangeless Male Nut for 1/16" OD Tubing
Qty/Pkg: EACH
Catalog No. 50-008-04598 Supplier Analytical Sales & Services Supplier No. 59230
1/4-28 PEEK Flangeless Male Nut for 1/16" OD Tubing
Qty/Pkg: EACH