Learn More
Analytical Sales & Services PEEK Guard Holder. 20mm, Direct Connect to Cartridges.
Catalog No. 5000804988
Encompass
Advantage™ PEEK Guard Holder. 20mm, Direct Connect to Cartridges.
Catalog No. 50-008-04988 Supplier Analytical Sales & Services Supplier No. ADVGUARDFM
Advantage™ PEEK Guard Holder. 20mm, Direct Connect to Cartridges.