Learn More
IDEX Health & Science Upchurch PEEK Manifold Assembly 9 Port, for 1/8" OD
Catalog No. NC1762169
Encompass_Preferred
PEEK Manifold Assembly can be applied for a multi-Ports mixing chamber, combining several fluid streams into one or one stream split to several outlets. It can also be used as a gas sparging splitting unions or sample injection onto multi-well plates or a multiple direction flow path union. The manifold is manufactured from biocompatible PEEK giving it excellent chemical resistance. It has 9 Ports and is for 1/8"OD tubing. This assembly comes with Super Flangeless Fittings (9) P-331 and (9) P-359.
Catalog No. NC1762169 Supplier IDEX Health & Science Upchurch Supplier No. P190
PEEK Manifold Assembly can be applied for a multi-Ports mixing chamber, combining several fluid streams into one or one stream split to several outlets. It can also be used as a gas sparging splitting unions or sample injection onto multi-well plates or a multiple direction flow path union. The manifold is manufactured from biocompatible PEEK giving it excellent chemical resistance. It has 9 Ports and is for 1/8"OD tubing. This assembly comes with Super Flangeless Fittings (9) P-331 and (9) P-359.