Learn More
Analytical Sales & Services PEEK MicroTight Adapter, 1/16" to 1/32" Tubing
Catalog No. 5000805045
Encompass
PEEK Micro Tight Adapter Assembly 10-32 to 6-32 Includes: (1) 55120R (1) 55126S Fittings and (1) 59554 Gauge Plug
Qty/Pkg: EACH
Catalog No. 50-008-05045 Supplier Analytical Sales & Services Supplier No. 59881
PEEK Micro Tight Adapter Assembly 10-32 to 6-32 Includes: (1) 55120R (1) 55126S Fittings and (1) 59554 Gauge Plug
Qty/Pkg: EACH