Learn More
Analytical Sales & Services PEEK MicroTight Adapter, 1/16" to 360µm Tubing
Catalog No. 5000804705
Encompass
PEEK Micro Tight Adapter Assembly, 10-32 to 6-32\n\nInlcludes: (1) 55120B Fitting, (1) 55124 Fitting and (1) 59554 Gauge Plug\n\nQty/Pkg: EACH
Catalog No. 50-008-04705 Supplier Analytical Sales & Services Supplier No. 59882
PEEK Micro Tight Adapter Assembly, 10-32 to 6-32\n\nInlcludes: (1) 55120B Fitting, (1) 55124 Fitting and (1) 59554 Gauge Plug\n\nQty/Pkg: EACH