Learn More
Analytical Sales & Services PEEK MicroTight Adapter, ZDV, 1/16" to Microsleeves
Catalog No. 5000804521
Encompass
PEEK MicroTight ZDV Adapter, 1/16" OD to MicroTight Sleeves, Includes; (1) 66514-1 (1) 55125 Fitting and (1) 59554 Gauge Plug\n\nQty/Pkg: EACH
Catalog No. 50-008-04521 Supplier Analytical Sales & Services Supplier No. 59770
Add to Cart
Add to Cart
PEEK MicroTight ZDV Adapter, 1/16" OD to MicroTight Sleeves, Includes; (1) 66514-1 (1) 55125 Fitting and (1) 59554 Gauge Plug\n\nQty/Pkg: EACH